missing translation for 'onlineSavingsMsg'
Learn More

Dihydromyrcenol, 99.1%, MP Biomedicals™

Product Code. 11426672 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 11426672

Brand: MP Biomedicals 0215090625

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 18479-58-8
Color Colorless
Molecular Formula C10H20O
MDL Number MFCD00004474
Quantity 25 g
Synonym dihydromyrcenol, 2,6-dimethyl-7-octen-2-ol, 7-octen-2-ol, 2,6-dimethyl, 1,1,5-trimethyl-6-heptenol, 1-2,6-dimethyloct-7-en-2-ol, dsstox_cid_9317, myrcenol, 6,10-dihydro, dsstox_rid_78761
InChI Key XSNQECSCDATQEL-UHFFFAOYSA-N
SMILES CC(CCCC(C)(C)O)C=C
IUPAC Name 2,6-dimethyloct-7-en-2-ol
Molecular Weight (g/mol) 156.269
PubChem CID 29096
ChEBI CHEBI:87528
Formula Weight 156.27
Physical Form Liquid
Chemical Name or Material Dihydromyrcenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.