Learn More
Dihydrochelerythrine, MedChemExpress
Product Code. 16479178
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16479178
Brand: Medchem Express HYN0903
MedChemExpress Dihydrochelerythrine is a natural compound isolated from the leaves of Macleaya microcarpa; has antifungal activity.
Dihydrochelerythrine is a natural compound isolated from the leaves of Macleaya microcarpa
Specifications
Chemical Name or Material Dihydrochelerythrine
Physical Form Solid
CAS 6880-91-7
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H19NO4
For Use With (Application) COVID-19-immunoregulation
Synonym 12,13-Dihydrochelerythrine
Solubility Information DMSO : 20.83 mg/mL (59.62 mM; Need ultrasonic)
SMILES CN1C(C2=CC(OCO3)=C3C=C2C=C4)=C4C5=CC=C(OC)C(OC)=C5C1
Molecular Weight (g/mol) 349.38
Formula Weight 349.38
Percent Purity 98.04%
Grade Research