missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Finasteride, TRC

Product Code. 18010430
Change view
Click to view available options
Quantity:
25 mg
50 mg
500 mg
Unit Size:
25mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18010430 50 mg 50mg
17919639 25 mg 25mg
18073430 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18010430 Supplier Toronto Research Chemicals Supplier No. TRCD44897550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98319-24-5
Molecular Formula C23 H38 N2 O2
Molecular Weight (g/mol) 374.56
Synonym N-(1,1-Dimethylethyl)-3-oxo-4-aza-5alpha-androstane-17beta-carboxamide, Finasteride Imp. A (EP), Dihydrofinasteride, 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-(1,1-dimethylethyl)hexadecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrostane-17-carboxamide, N-(1,1-dimethylethyl)-3-oxo-, (5α,17β)-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-(1,1-Dimethylethyl)hexadecahydro-4a,6a-dimethyl-2-oxo-1H-indeno[5,4-f]quinoline-7-carboxamide, 1,2-Dihydrofinasteride, 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-(1,1-dimethylethyl)hexadecahydro-4a,6a-dimethyl-2-oxo-, [4aR-(4aα,4bβ,6aα,7α,9aβ,9bα,11aβ)]-, 3-Oxo-4-aza-5α-androstan-17β-carboxylic acid N-(tert-butyl)amide, N-(tert-Butyl)-3-oxo-4-aza-5α-androstane-17-carboxamide, N-(tert-Butyl)-3-oxo-4-aza-5α-androstane-17β-carboxamide
IUPAC Name (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,8,9,9b,10,11-tetradecahydroindeno[5,4-f]quinoline-1-carboxamide
SMILES CC(C)(C)NC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H38N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h14-18H,6-13H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,18+,22-,23+/m0/s1
Formula Weight 374.2933
Chemical Name or Material Dihydrofinasteride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.