Learn More
Karl Hecht™ Digital Clinical Thermometer with Signal
Click to view available options
Temperature Range (Metric):
32°C to 42°C
Unit Size:
Each
Description
- Electronic.
- Waterproof.
- LCD display.
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| For Use With (Application) | Suitable for oral, rectal and axillary measurement |
| Measurement Time | Approx. 1 min. at room temperature |
| Product Type | Digital Clinical Thermometer with Signal |
| Max. Temperature (Metric) | 42°C |
| Min. Temperature (Metric) | 32°C |
| Temperature Range (Metric) | 32°C to 42°C |
| Battery Life | Approx. 250 hr. |
| Battery Type | LR 41 |
| Accuracy | ±0.1°C |
| Show More |