Learn More
Cole-Parmer™ Digi-Sense™ Power Adapter for Temperature Datalogging Meters
Shop All Cole Parmer Products
Click to view available options
Voltage:
115/240 V
Unit Size:
Each
Description
- For use with Oakton temperature datalogging meters.
- Powers the meter with out the need for batteries.
- If you are a 24-hour lab or heavy use laboratory, you need the meter plugged into an electrical source.
Specifications
Specifications
| For Use With (Equipment) | Temperature Datalogging Meters |
| Voltage | 115/240 V |