Learn More
Thermo Scientific Chemicals Diethylcarbamazine
Product Code. 16514424 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16514424 25 g 25g
16504424 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16514424 Supplier Thermo Scientific Chemicals Supplier No. 464510250
Request Bulk or Custom Format
CAS: 90-89-1 | C10H21N3O | 199.30 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90-89-1
Molecular Formula C10H21N3O
Molecular Weight (g/mol) 199.30
InChI Key RCKMWOKWVGPNJF-UHFFFAOYSA-N
IUPAC Name N,N-diethyl-4-methylpiperazine-1-carboxamide
SMILES CCN(CC)C(=O)N1CCN(C)CC1
Specifications
Melting Point 49.0°C to 51.0°C
Quantity 25 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 199.29
Chemical Name or Material Diethylcarbamazine
compliance-icons
Product Identifier
  • Diethylcarbamazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Diethylcarbamazine