Learn More
Diethyl oxalacetate sodium salt, 95%
Product Code. p-7025807
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11487024 100 g 100g
11497024 500 g 500g
2 options
Product Code. 11487024
Brand: Thermo Scientific Alfa Aesar B21486.22
Request Bulk or Custom Format
CAS: 40876-98-0 | C8H11NaO5 | 210.161 g/mol
Diethyl oxalacetate sodium salt is used in the preparation of oxaloacetic acid. It is also used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl oxalacetate sodium salt is used in the preparation of oxaloacetic acid. It is also used as an active pharmaceutical intermediate.
Solubility
Slightly soluble in water (1.2 g/L) (25°C).
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 40876-98-0
Molecular Formula C8H11NaO5
Molecular Weight (g/mol) 210.161
MDL Number MFCD00035571
InChI Key UJZUICGIJODKOS-UHFFFAOYSA-M
Synonym diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
PubChem CID 44134875
IUPAC Name sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate
SMILES CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]
Specifications
Melting Point ∼185°C
Quantity 100 g
Beilstein 4279609
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water (1.2g/L) (25°C).
Formula Weight 210.16
Percent Purity 95%
Chemical Name or Material Diethyl oxalacetate sodium salt