missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Isopropenyl Phosphate, TRC

Product Code. 30369914
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369914 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369914 Supplier Toronto Research Chemicals Supplier No. TRCD4960835G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Isopropenyl Phosphate
CAS 5954-28-9
Molecular Formula C7 H15 O4 P
Quantity 5 g
InChI Formula InChI=1S/C7H15O4P/c1-5-9-12(8,10-6-2)11-7(3)4/h3,5-6H2,1-2,4H3
SMILES CCOP(=O)(OCC)OC(=C)C
IUPAC Name diethyl prop-1-en-2-yl phosphate
Molecular Weight (g/mol) 194.165
Formula Weight 194.071
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.