Learn More
Diethyl acetylenedicarboxylate, 97%
Product Code. p-108177 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
Product Code. 10762491
Brand: Thermo Scientific Acros undefined
Request Bulk or Custom Format
CAS: 762-21-0 | C8H10O4 | 170.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 762-21-0
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.17
MDL Number MFCD00009186
InChI Key STRNXFOUBFLVIN-UHFFFAOYSA-N
Synonym diethyl acetylenedicarboxylate, 2-butynedioic acid, diethyl ester, diethyl 2-butynedioate, 1,4-diethyl but-2-ynedioate, acetylenedicarboxylic acid, diethyl ester, acetylenedicarboxylic acid diethyl ester, diethylacetylenedicarboxylate, bis-ethoxycarbonyl acetylene, diethyl acetylene dicarboxylate, di-ethyl acetylenedicarboxylate
PubChem CID 69803
IUPAC Name diethyl but-2-ynedioate
SMILES CCOC(=O)C#CC(=O)OCC
Specifications
Melting Point 1.0°C to 3.0°C
Color Yellow
Density 1.0600g/mL
Boiling Point 107.0°C to 110.0°C (11.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4410 to 1.4430
Quantity 25 g
Beilstein 02, 803
Fieser 01,244; 02,127
Specific Gravity 1.06
Solubility Information Solubility in water: .. Other solubilities: soluble in ethyl alcohol,ethyl ether,carbon tetr
Formula Weight 170.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl acetylenedicarboxylate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl acetylenedicarboxylate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.