Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Dieldrin |
| CAS | 60-57-1 |
| CAS Max % | ≤ 100% |
| Beilstein | 91396 |
| Color | Light Brown |
| Melting Point | 175°C to 176°C |
| Synonym | 1,2,3,4,10,10-Hexachloro-1,4,4a,5,6,7,8,8a-octahydro-6,7-epoxy-1,4:5,8-dimethanonaphthalene |
| Molecular Weight (g/mol) | 380.9 g/mol |
| Quantity | 100 mg |