Learn More
Dichloro(dimethylamino)phosphine, 98%
Product Code. 11357459
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357459 5 g 5g
11367459 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11357459 Supplier Thermo Scientific Alfa Aesar Supplier No. H27856.06
Request Bulk or Custom Format
CAS: 683-85-2 | C2H6Cl2NP | 145.951 g/mol
Dimethylphosphoramidous dichloride (N,N-dimethylamidodichlorophosphite) may be used in the synthesis of the following N,N-dimethylamidotetrachlorophosphorane, O-propylchloroformimino-N,N-dimethylamidochlorophosphate, bisarylsulphonyldiazadiphosphetidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethylphosphoramidous dichloride (N,N-dimethylamidodichlorophosphite) may be used in the synthesis of the following N,N-dimethylamidotetrachlorophosphorane, O-propylchloroformimino-N,N-dimethylamidochlorophosphate, bisarylsulphonyldiazadiphosphetidines.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, heat.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 683-85-2
Molecular Formula C2H6Cl2NP
Molecular Weight (g/mol) 145.951
MDL Number MFCD00013613
InChI Key XPWWDZRSNFSLRQ-UHFFFAOYSA-N
Synonym dimethylphosphoramidous dichloride, dichloro dimethylamino phosphine, phosphoramidous dichloride, dimethyl, dimethylamino dichlorophosphine, dimethylphosphoramidousdichloride, me2npcl2, dimethylaminodichlorphosphine, dimethylaminodichlorophosphine, ch3 2npcl2, dichlorophosphanyl dimethylamine
PubChem CID 136483
IUPAC Name N-dichlorophosphanyl-N-methylmethanamine
SMILES CN(C)P(Cl)Cl
Specifications
Density 1.271
Boiling Point 150°C
Flash Point −23°C (−9°F)
Linear Formula (CH3)2NPCl2
Quantity 5 g
UN Number UN2924
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 145.96
Percent Purity 98%
Chemical Name or Material Dichloro(dimethylamino)phosphine