Learn More
Dibenzyl phthalate, 97%
Product Code. 11371787
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371787 25 g 25g
11361787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11371787 Supplier Thermo Scientific Alfa Aesar Supplier No. L04985.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 523-31-9 | C22H18O4 | 346.382 g/mol
Dibenzyl phthalate is an plasticizer used in toys that was found to exhibit potential estrogen agonist/ antagonist properties. It is used in PVC, paints, acrylic coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dibenzyl phthalate is an plasticizer used in toys that was found to exhibit potential estrogen agonist/ antagonist properties. It is used in PVC, paints, acrylic coatings.
Solubility
Insoluble in water.
Notes
Incompatible materials are reducing agents. Strong acids. Strong bases and Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 523-31-9
Molecular Formula C22H18O4
Molecular Weight (g/mol) 346.382
MDL Number MFCD00014437
InChI Key UCVPKAZCQPRWAY-UHFFFAOYSA-N
Synonym dibenzyl phthalate, benzyl phthalate, 1,2-benzenedicarboxylic acid, bis phenylmethyl ester, phthalic acid, dibenzyl ester, dibenzylphthalate, 1, bis phenylmethyl ester, acmc-1b21b, phthalic acid dibenzyl ester, phthalic acid, bis-benzylester, dibenzyl phthalate, analytical standard
PubChem CID 220773
IUPAC Name dibenzyl benzene-1,2-dicarboxylate
SMILES C1=CC=C(C=C1)COC(=O)C2=CC=CC=C2C(=O)OCC3=CC=CC=C3
Specifications
Melting Point 40°C to 42°C
Boiling Point 276°C to 278°C (15 mmHg)
Quantity 25 g
Beilstein 2338250
Solubility Information Insoluble in water.
Formula Weight 346.39
Percent Purity 97%
Chemical Name or Material Dibenzyl phthalate