Learn More
Dibenz[a,h]anthracene-d14, CDN
Product Code. 30327425
Change view
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327425 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30327425 Supplier CDN Supplier No. CDND05200.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dibenzo(a,h)anthracene D14
CAS 13250-98-1
Molecular Formula C22 2H14
Quantity 0.01 g
Synonym Dibenz[a,h]anthracene-1,2,3,4,5,6,7,8,9,10,11,12,13,14-d14, Dibenz[a,h]anthracene-d14 (7 CI,8 CI,9 CI), Dibenzo[a,h]anthracene D14, Dibenzo(a,h)anthracene D14
InChI Formula InChI=1 S/C22H14/c1-3-7-19-15(5-1)9-11-17-14-22-18(13-21(17)19)12-10-16-6-2-4-8-20(16)22/h1-14 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D,14 D
SMILES [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c([2 H])c([2 H])c3c([2 H])c4c(c([2 H])c([2 H])c5c([2 H])c([2 H])c([2 H])c([2 H])c45)c([2 H])c23
IUPAC Name 1,2,3,4,5,6,7,8,9,10,11,12,13,14-tetradecadeuterionaphtho[1,2-b]phenanthrene
Molecular Weight (g/mol) 292.43
Formula Weight 292.1974 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity