missing translation for 'onlineSavingsMsg'
Learn More

Dianthramine, TRC

Product Code. 30366947
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366947 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366947 Supplier Toronto Research Chemicals Supplier No. TRCD2097355MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Dianthramine
CAS 136945-65-8
Molecular Formula C14 H11 N O6
Quantity 5 mg
InChI Formula InChI=1S/C14H11NO6/c16-7-1-3-9(13(18)19)11(5-7)15-12-6-8(17)2-4-10(12)14(20)21/h1-6,15-17H,(H,18,19)(H,20,21)
SMILES OC(=O)c1ccc(O)cc1Nc2cc(O)ccc2C(=O)O
IUPAC Name 2-(2-carboxy-5-hydroxyanilino)-4-hydroxybenzoic acid
Molecular Weight (g/mol) 289.24
Formula Weight 289.059
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.