missing translation for 'onlineSavingsMsg'
Learn More

Diallyl sulfide, 98%

Product Code. p-7045504
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11363106

Brand: Thermo Scientific Alfa Aesar L10971.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-88-1 | C6H10S | 114.21 g/mol

Diallyl sulfide is a modulator of drug metabolizing enzyme P450 system and inducer of the phase II detoxifying enzyme GST. Diallyl sulfide is also an inhibitor of CYP2E1 and believed to prevent chemically-induced carcinogenesis in many tissues. It is also used as a food additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diallyl sulfide is a modulator of drug metabolizing enzyme P450 system and inducer of the phase II detoxifying enzyme GST. Diallyl sulfide is also an inhibitor of CYP2E1 and believed to prevent chemically-induced carcinogenesis in many tissues. It is also used as a food additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in alcohol, chloroform, ether, and carbon tetrachloride. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-88-1
Molecular Formula C6H10S
Molecular Weight (g/mol) 114.21
MDL Number MFCD00008658
InChI Key UBJVUCKUDDKUJF-UHFFFAOYSA-N
Synonym diallyl sulfide, allyl sulfide, oil garlic, diallylsulfide, thioallyl ether, 1-propene, 3,3'-thiobis, allyl monosulfide, diallyl thioether, diallyl monosulfide, diallyl sulphide
PubChem CID 11617
ChEBI CHEBI:4489
SMILES C=CCSCC=C

Specifications

Melting Point -83°C
Density 0.889
Boiling Point 137°C to 138°C
Flash Point 23°C (73°F)
Odor Unpleasant
Linear Formula CH2=CHCH2SCH2CH=CH2
Refractive Index 1.4895
Quantity 25 g
UN Number UN1993
Beilstein 1736016
Merck Index 14,297
Solubility Information Soluble in alcohol,chloroform,ether,and carbon tetrachloride. Insoluble in water.
IUPAC Name 3-prop-2-enylsulfanylprop-1-ene
Formula Weight 114.21
Percent Purity 98%
Chemical Name or Material Diallyl sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.