missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHRS2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16518331
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16518331 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16518331 Supplier Invitrogen™ Supplier No. PA592982

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MLSAVARGYQ GWFHPCARLS VRMSSTGIDR KGVLANRVAV VTGSTSGIGF AIARRLARDG AHVVISSRKQ QNVDRAMAKL QGEGLSVAGI VCHVGKAEDR EQLVAKALEH CGGVDFLVCS AGVNPLVGST LGTSEQIWDK ILSVNVKSPA LLLSQLLPYM ENRRGAVILV SSIAAYNPVV ALGVYNVSKT ALLGLTRTLA LELAPKDIRV NCVVPGIIKT DFSKVVRIGF MGMSLSGRTS RNIISCRGLG SQRTVQESCP SCALQMPATS TGRTLRWQAT PLGSERSGGG CVAVVPGPGA; Positive Samples: HepG2, SKOV3, MCF7, BT-474, 22Rv1, Mouse liver, Mouse heart, Mouse bone marrow, Rat liver; Cellular Location: Mitochondrion matrix, Nucleus.

DHRS2 displays NADPH-dependent dicarbonyl reductase activity in vitro with 3,4-Hexanedione, 2,3-Heptanedione and 1-Phenyl-1,2-propanedione as substrates. DHRS2 do not reductase activity is displayed in vitro with steroids, retinoids and sugars as substrates. This protein may inhibit cell replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DHRS2
Gene Accession No. Q13268
Gene Alias 5430405K24Rik; dehydrogenase/reductase (SDR family) member 2; dehydrogenase/reductase 2; dehydrogenase/reductase member 2; dehydrogenase/reductase SDR family member 2, mitochondrial; Dhrs2; Dicarbonyl reductase HEP27; Hep27; Protein D; SDR family member; SDR25C1; Short chain dehydrogenase/reductase family 25C member 1; short chain dehydrogenase/reductase family 25C, member 1; short-chain alcohol dehydrogenase family member
Gene Symbols DHRS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human DHRS2 (NP_8789121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10202, 691464, 71412
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.