missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DHRS1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15719380 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15719380 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15719380 Supplier Invitrogen™ Supplier No. PA551420

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NKAFWETPAS MWDDINNVGL RGHYFCSVYG ARLMVPAGQG LIVVISSPGS LQYMFNVPYG VGKAACDKLA ADCAHELRRH GVSCVSLWPG IVQTELLKEH MAKEEVLQDP VLKQFKSAFS SAETTELSGK CVVALATDPN ILSLSGKVLP SCDLARRYGL RDVDGRPVQD YLSLSSVLSH VSGLGWLASY LPSFLRVPKW Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 83%.

This gene encodes a member of the short-chain dehydrogenases/reductases (SDR) family. The encoded enzyme contains a conserved catalytic domain and likely functions as an oxidoreductase. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dhrs1
Gene Accession No. Q96LJ7
Gene Alias 1110029G07Rik; AW112170; D14ertd484e; dehydrogenase/reductase (SDR family) member 1; dehydrogenase/reductase 1; dehydrogenase/reductase SDR family member 1; DHRS1; SDR19C1; Short chain dehydrogenase/reductase family 19C member 1; short chain dehydrogenase/reductase family 19C, member 1
Gene Symbols Dhrs1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHRS1. Recombinant protein control fragment (Product #RP-88880).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.