missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGAT2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05769MFV500
This item is not returnable.
View return policy
Description
DGAT2 Polyclonal antibody specifically detects DGAT2 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
| DGAT2 | |
| Polyclonal | |
| Flow Cytometry, Flow (Intracellular) | |
| diacylglycerol O-acyltransferase 2, diacylglycerol O-acyltransferase homolog 2, diacylglycerol O-acyltransferase homolog 2 (mouse), diacylglycerol O-acyltransferase-like protein 2, Diglyceride acyltransferase 2, DKFZp686A15125, EC 2.3.1, EC 2.3.1.20, GS1999FULL | |
| Partial recombinant protein made to an internal portion of the human DGAT2 protein (amino acids 268-377) [UniProt Q96PD7] | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 84649 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction