missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGAT1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-41487R
This item is not returnable.
View return policy
Description
DGAT1 Polyclonal specifically detects DGAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoblotting.
Specifications
| DGAT1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoblotting | |
| O75907 | |
| DGAT1 | |
| A synthetic peptide made to an internal region (within residues 200-300) of human DGAT1. [Swiss-Prot# O75907] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, bovine, Zebrafish, and rat. Predicted to react with primate, sheep, and goat based on 100% sequence homology. | |
| Store at 4C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ACAT-related gene product 1, AGRP1, ARGP1acyl coenzyme A:cholesterol acyltransferase related gene 1, DGATACAT related gene product 1, diacylglycerol O-acyltransferase 1, diacylglycerol O-acyltransferase homolog 1, diacylglycerol O-acyltransferase homolog 1 (mouse), Diglyceride acyltransferase, EC 2.3.1, EC 2.3.1.20 | |
| Rabbit | |
| 55 kDa | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 8694 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction