missing translation for 'onlineSavingsMsg'
Learn More

DG051, MedChemExpress

Product Code. p-200098053
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16491099

Brand: Medchem Express HY10825

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DG051 is a potent leukotriene A4 hydrolase inhibitor of leukotriene B4 biosynthesis in the enzyme assay with an IC50=47 nM.

DG051 is a potent leukotriene A4 hydrolase inhibitor of leukotriene B4 biosynthesis in the enzyme assay with an IC50=47 nM.

Specifications

Chemical Name or Material DG051
CAS 929915-58-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H25Cl2NO4
Quantity 1 mg
Solubility Information DMSO : ≥ 325 mg/mL (762.32 mM)
SMILES Cl[H].O=C(O)CCCN1CCC[C@H]1COC(C=C2)=CC=C2OC3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 426.33
Formula Weight 426.33
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.