missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexchlorpheniramine maleate, MedChemExpress
Description
Dexchlorpheniramine maleate is an antihistamine, with anticholinergic properties, used to treat allergic conditions.
Specifications
Specifications
| Chemical Name or Material | Dexchlorpheniramine maleate |
| CAS | 2438-32-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H23ClN2O4 |
| Quantity | 200 mg |
| Synonym | S-(+)-Chlorpheniramine maleate salt |
| Solubility Information | DMSO : ≥ 100 mg/mL (255.85 mM) ∣H2O : ≥ 100 mg/mL (255.85 mM) |
| SMILES | O=C(O)/C=C\C(O)=O.CN(C)CC[C@H](C1=NC=CC=C1)C2=CC=C(Cl)C=C2 |
| Molecular Weight (g/mol) | 390.86 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?