missing translation for 'onlineSavingsMsg'
Learn More

Detomidine Hydrochloride, TRC

Product Code. 18053640
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18053640 10 mg 10mg
30135842 1 g 1g
18022040 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18053640 Supplier Toronto Research Chemicals Supplier No. TRCD29797510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90038-01-0
Molecular Formula C12 H14 N2 . Cl H
Molecular Weight (g/mol) 222.71
Synonym Detomidine hydrochloride, 4-(2,3-Dimethylbenzyl)-1H-imidazole hydrochloride, 1H-Imidazole, 4-[(2,3-dimethylphenyl)methyl]-, monohydrochloride, Detomidine hydrochloride, Domosedan, Dormosedan, MPV 253AII
IUPAC Name 4-[(2,3-dimethylphenyl)methyl]-1H-imidazole;hydrochloride
SMILES Cl.Cc1cccc(Cc2c[nH]cn2)c1C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H14N2.ClH/c1-9-4-3-5-11(10(9)2)6-12-7-13-8-14-12;/h3-5,7-8H,6H2,1-2H3,(H,13,14);1H
Formula Weight 222.0924
Chemical Name or Material Detomidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.