missing translation for 'onlineSavingsMsg'
Learn More

Desmosteryl Sulfate Sodium Salt, TRC

Product Code. 30371118
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371118 10 mg 10mg
30374220 5 mg 5mg
30370306 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30371118 Supplier Toronto Research Chemicals Supplier No. TRCD43250210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmosteryl Sulfate Sodium Salt
Analyte or Component Names Desmosteryl Sulfate Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C27 H43 Na O4 S
Synonym Sodium (3S,8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylhept-5-en-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl sulfate
InChI Formula InChI=1S/C27H44O4S.Na/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(31-32(28,29)30)13-15-26(20,4)25(22)14-16-27(23,24)5;/h7,9,19,21-25H,6,8,10-17H2,1-5H3,(H,28,29,30);/q;+1/p-1/t19-,21+,22+,23-,24+,25+,26+,27-;/m1./s1
Solution Type Neat
SMILES C[C@H](CCC=C(C)C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OS(=O)(=O)O[Na]
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylhept-5-en-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxysulfonyloxysodium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 486.683
Quantity 10 mg
Formula Weight 486.278
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.