missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmin Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00 EUR - 349.00 EUR
Specifications
| Antigen | Desmin |
|---|---|
| Clone | 2B1A11 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235905
|
Proteintech
67793-1-IG-20UL |
20 μL |
145.00 EUR 129.00 EUR / 20µL Save 16.00 EUR 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236736
|
Proteintech
67793-1-IG-150UL |
150 μL |
410.00 EUR 349.00 EUR / 150µL Save 61.00 EUR 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Desmin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Desmin. This antibody recognizes human, mouse, pig antigen. The Desmin antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| Desmin | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 1674 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2B1A11 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Pig | |
| CMD1I; CSM1; CSM2; FLJ12025; FLJ39719; FLJ41013; FLJ41793 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title