missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmethylchlorphenamine Maleate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Desmethylchlorphenamine Maleate |
| CAS | 22630-25-7 |
| Analyte or Component Names | Desmethylchlorphenamine Maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H17 Cl N2 . C4 H4 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (3RS)-3-(4-Chloro-phenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine Maleate, Chlorphenamine Maleate Imp. C (EP) as Maleate, Desmethylchlorphenamine Maleate, Chlorphenamine Maleate Imp. C (EP), Chlorphenamine Imp. C (EP), Chlorpheniramine USP Related Compound C, Chlorpheniramine USP RC C, Chlorpheniramine Related Compound C as Maleate, Chlorphenamine Maleate Impurity C as Maleate, Chlorphenamine Impurity C as Maleate |
| InChI Formula | InChI=1S/C15H17ClN2.C4H4O4/c1-17-11-9-14(15-4-2-3-10-18-15)12-5-7-13(16)8-6-12;5-3(6)1-2-4(7)8/h2-8,10,14,17H,9,11H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| Solution Type | Neat |
| SMILES | CNCCC(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?