missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Sibutramine, Hydrochloride Salt, TRC

Product Code. 30135479
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135479 10 mg 10mg
30137867 5 mg 5mg
30012222 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135479 Supplier Toronto Research Chemicals Supplier No. TRCD29217010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84467-94-7
Molecular Formula C16 H24 Cl N . Cl H
Molecular Weight (g/mol) 302.28
Synonym N-Monodesmethylsibutramine hydrochloride, Cyclobutanemethanamine, 1-(4-chlorophenyl)-N-methyl-α-(2-methylpropyl)-, hydrochloride (1:1), Cyclobutanemethanamine, 1-(4-chlorophenyl)-N-methyl-α-(2-methylpropyl)-, hydrochloride (9CI), BTS 54-354, Desmethylsibutramine hydrochloride, (1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine hydrochloride
IUPAC Name 1-[1-(4-chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine;hydrochloride
SMILES Cl.CNC(CC(C)C)C1(CCC1)c2ccc(Cl)cc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H24ClN.ClH/c1-12(2)11-15(18-3)16(9-4-10-16)13-5-7-14(17)8-6-13;/h5-8,12,15,18H,4,9-11H2,1-3H3;1H
Formula Weight 301.1364
Percent Purity >95%
Chemical Name or Material Desmethylsibutramine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.