missing translation for 'onlineSavingsMsg'
Learn More

Deshydroxyethoxy Ticagrelor, TRC

Product Code. 30004670
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004670 5 mg 5mg
30005511 10 mg 10mg
30006054 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004670 Supplier Toronto Research Chemicals Supplier No. TRCD2900455MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220347-05-7
Molecular Formula C21 H24 F2 N6 O3 S
Molecular Weight (g/mol) 478.52
Synonym (1S,2R,3S,4R)-4-[7-[[(1R,2S)-2-(3,4-Difluorophenyl)cyclopropyl]amino]-5-(propylthio)-3H-1,2,3-triazolo[4,5-d]pyrimidin-3-yl]-1,2,3-cyclopentanetriol, AR-C 124910XX
IUPAC Name (1S,2R,3S,4R)-4-[7-[[(1R,2S)-2-(3,4-difluorophenyl)cyclopropyl]amino]-5-propylsulfanyltriazolo[4,5-d]pyrimidin-3-yl]cyclopentane-1,2,3-triol
SMILES CCCSc1nc(N[C@@H]2C[C@H]2c3ccc(F)c(F)c3)c4nnn([C@@H]5C[C@H](O)[C@@H](O)[C@H]5O)c4n1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H24F2N6O3S/c1-2-5-33-21-25-19(24-13-7-10(13)9-3-4-11(22)12(23)6-9)16-20(26-21)29(28-27-16)14-8-15(30)18(32)17(14)31/h3-4,6,10,13-15,17-18,30-32H,2,5,7-8H2,1H3,(H,24,25,26)/t10-,13+,14+,15-,17-,18+/m0/s1
Formula Weight 478.1599
Percent Purity >95%
Chemical Name or Material Deshydroxyethoxy Ticagrelor
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.