missing translation for 'onlineSavingsMsg'
Learn More

Desfesoterodine, MedChemExpress

Product Code. 16497946
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497946 100 mg 100mg
16494107 50 mg 50mg
16488208 10 mg 10mg
16460648 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497946 Supplier Medchem Express Supplier No. HY76569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Desfesoterodine (PNU-200577) is a potent and selective muscarinic receptor (mAChR) antagonist with a KB and a pA2 of 0.84 nM and 9.14, respectively. Desfesoterodine is a major pharmacologically active metabolite of Tolterodine (PNU-200583; HY-A0024) and Fesoterodine (HY-70053). Desfesoterodine improves cerebral infarction induced detrusor overactivity in rats.

Desfesoterodine (PNU-200577) is a potent and selective muscarinic receptor (mAChR) antagonist with a KB and a pA2 of 0.84 nM and 9.14, respectively. Desfesoterodine is a major pharmacologically active metabolite of Tolterodine (PNU-200583

Specifications

Chemical Name or Material Desfesoterodine
CAS 207679-81-0
Color White
Purity Grade Notes Research
Molecular Formula C22H31NO2
Quantity 100 mg
Synonym PNU-200577 5-Hydroxymethyl Tolterodine
Solubility Information DMSO : ≥ 100 mg/mL (292.83 mM)
SMILES OC1=C(C=C(CO)C=C1)[C@@H](C2=CC=CC=C2)CCN(C(C)C)C(C)C
Molecular Weight (g/mol) 341.49
Formula Weight 341.49
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.