missing translation for 'onlineSavingsMsg'
Learn More

Descarbamoyl Cefuroxime, TRC

Product Code. 30138415
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138415 5 mg 5mg
30007841 10 mg 10mg
30008628 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138415 Supplier Toronto Research Chemicals Supplier No. TRCD2888305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56271-94-4
Molecular Formula C15 H15 N3 O7 S
Molecular Weight (g/mol) 381.36
Synonym Descarbamoylcefuroxime, Cefuroxime Sodium Imp. A, 640/2, Ph Eur Cefuroxime Sodium Impurity A
IUPAC Name (6R,7R)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-3-(hydroxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)CO)\c3occc3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H15N3O7S/c1-24-17-9(8-3-2-4-25-8)12(20)16-10-13(21)18-11(15(22)23)7(5-19)6-26-14(10)18/h2-4,10,14,19H,5-6H2,1H3,(H,16,20)(H,22,23)/b17-9-/t10-,14-/m1/s1
Formula Weight 381.0631
Chemical Name or Material Descarbamoyl Cefuroxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.