missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEN2D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113487
This item is not returnable.
View return policy
Description
DEN2D Polyclonal Antibody for ELISA, IHC (P), Western Blot
Specifications
| DEN2D | |
| Polyclonal | |
| Unconjugated | |
| DENND2D | |
| 2010308M01Rik; AI451421; DENN domain containing 2D; DENN domain-containing protein 2D; DENN/MADD domain containing 2D; DENND2D; RGD1305120; RP5-1180E21.2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 79961 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H6A0 | |
| DENND2D | |
| Recombinant Human DENN domain-containing protein 2D protein (1-233AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction