Learn More
Emma Safety Footwear Dempo Safety Shoes
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Sanitized
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Color | Black |
| Design | Dempo |
| Liner Material | Hydro-Tec™ Sanitized Silver lined tongue |
| Material | Leather |
| Size | 45 |
| Toe Material | Steel |