Learn More
Demeclocycline hydrochloride, MedChemExpress
Product Code. 16481876
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16481876
Brand: Medchem Express HY17560
MedChemExpress Demeclocycline hydrochloride is a tetracycline antibiotic; is an antibiotic in the treatment of Lyme disease, acne, and bronchitis.
Demeclocycline hydrochloride is a tetracycline antibiotic
Specifications
Chemical Name or Material Demeclocycline hydrochloride
CAS 64-73-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H22Cl2N2O8
Quantity 500 mg
Solubility Information DMSO : 100 mg/mL (199.48 mM; Need ultrasonic) ∣H2O : 20 mg/mL (39.90 mM; Need ultrasonic and warming)
SMILES O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])[C@H](O)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4Cl)N.Cl
Molecular Weight (g/mol) 501.31
Formula Weight 501.31
Percent Purity 95.09%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation