missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deltarasin, MedChemExpress
Description
Deltarasin is an inhibitor of KRAS-PDEδ interaction with Kd of 38 nM for binding to purified PDEδ.
Specifications
Specifications
| Chemical Name or Material | Deltarasin |
| CAS | 1440898-61-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C40H37N5O |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 42 mg/mL (69.57 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | C1(C2=CC=C(OC[C@@H](N3C4=CC=CC=C4N=C3C5=CC=CC=C5)C6CCNCC6)C=C2)=NC7=CC=CC=C7N1CC8=CC=CC=C8 |
| Molecular Weight (g/mol) | 603.75 |
| Formula Weight | 603.75 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?