missing translation for 'onlineSavingsMsg'
Learn More

Delpazolid, MedChemExpress

Product Code. p-200097544
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16469486

Brand: Medchem Express HY100180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Delpazolid is a novel oxazolidinone antibiotic agent which can inhibit the growth of MSSA and MRSA with a MIC90 of 2 μg/mL for both of them.

Delpazolid is a novel oxazolidinone antibiotic agent which can inhibit the growth of MSSA and MRSA with a MIC90 of 2 μg/mL for both of them.

Specifications

Chemical Name or Material Delpazolid
Physical Form Solid
CAS 1219707-39-7
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H17FN4O3
For Use With (Application) COVID-19-immunoregulation
Synonym LCB01-0371
Solubility Information DMSO : 30 mg/mL (97.30 mM; Need ultrasonic and warming)
SMILES O=C1O[C@@H](CO)CN1C2=CC=C(N3C=NN(C)CC3)C(F)=C2
Molecular Weight (g/mol) 308.31
Formula Weight 308.31
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.