missing translation for 'onlineSavingsMsg'
Learn More

Defluoro Levofloxacin, TRC

Product Code. 30354521
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354521 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354521 Supplier Toronto Research Chemicals Supplier No. TRCD2290201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desfluorolevofloxacin
CAS 117620-85-6
Molecular Formula C18 H21 N3 O4
Quantity 1 mg
Synonym 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-, (3S)-, 7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-, (S)-, (3S)-2,3-Dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, (S)-2,3-Dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de][1,4]benzoxazine-6-carboxylic acid, Desfluorolevofloxacin
InChI Formula InChI=1S/C18H21N3O4/c1-11-10-25-17-14(20-7-5-19(2)6-8-20)4-3-12-15(17)21(11)9-13(16(12)22)18(23)24/h3-4,9,11H,5-8,10H2,1-2H3,(H,23,24)/t11-/m0/s1
SMILES C[C@H]1COc2c(ccc3C(=O)C(=CN1c23)C(=O)O)N4CCN(C)CC4
Molecular Weight (g/mol) 343.38
Formula Weight 343.1532
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.