Learn More
GA International Deep-Freeze Labels for Thermal-Transfer Barcode Printers
Product Code. 17820738
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
Brown
Green
Lavender
Lemon
Mint
Orange
Pink
Red
Silver
Yellow
Dimensions (LxW):
22 x 15 mm
25.4 x 12.7 mm
31.8 x 22.2 mm
38.1 x 19.1 mm
76.2 x 25.4 mm
Unit Size:
Pack of 1000
Pack of 3000
Product Code. Color Dimensions (LxW) unitSize
17820738 Yellow 31.8 x 22.2 mm Pack of 1000
17801288 Orange 38.1 x 19.1 mm Pack of 1000
17821008 Yellow 22 x 15 mm Pack of 3000
17889287 Blue 76.2 x 25.4 mm Pack of 1000
17850898 Brown 76.2 x 25.4 mm Pack of 1000
17810608 Green 76.2 x 25.4 mm Pack of 1000
17841188 Lavender 76.2 x 25.4 mm Pack of 1000
17860028 Orange 76.2 x 25.4 mm Pack of 1000
17880698 Pink 76.2 x 25.4 mm Pack of 1000
17840608 Red 76.2 x 25.4 mm Pack of 1000
17869437 Silver 76.2 x 25.4 mm Pack of 1000
17889537 Yellow 76.2 x 25.4 mm Pack of 1000
17829137 Blue 25.4 x 12.7 mm Pack of 1000
17850318 Green 25.4 x 12.7 mm Pack of 1000
17860888 Lemon 25.4 x 12.7 mm Pack of 1000
17849817 Pink 25.4 x 12.7 mm Pack of 1000
17859327 Silver 25.4 x 12.7 mm Pack of 1000
17879427 Yellow 25.4 x 12.7 mm Pack of 1000
17820438 Green 31.8 x 22.2 mm Pack of 1000
17820158 Mint 31.8 x 22.2 mm Pack of 1000
17859657 Orange 31.8 x 22.2 mm Pack of 1000
17801218 - 31.8 x 22.2 mm Pack of 1000
22 options
Product Code. 17820738
Brand: GA International FJT9C11YE
Only null left
Add to basket
Add to basket
Permanent deep-freeze labels for storage of samples in lab refrigerators, ultra-low temperature freezers, dry ice transport and general lab use.
  • Ideal for long-term storage in deep-freeze conditions (-80°C)
  • No curing time, can be stored in freezers immediately after application
  • No additional lamination necessary
  • Labels and printout resist spraying with alcohols and other chemicals
  • Printable with all major thermal-transfer printer brands
  • FDA Compliance: FDA 21 CFR 175.105 for indirect food contact
  • RoHS compliant
  • Waterproof and tearproof
  • Adhere to moist and wet surfaces
  • Resist spraying with alcohols and withstand multiple freeze-thaw cycles
Specifications
Color Yellow
Finish Matte
Format Roll
Material Thermoplastic Film
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Labels per Row 1 Label
Quantity 1000/Pk.
Shape Rectangle
Temperature Range (English) -112°F to +230°F
Temperature Range (Metric) -80°C to +110°C
Dimensions (LxW) 31.8 x 22.2 mm
Length (English) 0.88 in.
Length (Metric) 22.2 mm
Width (English) 1.25 in.
Width (Metric) 31.8 mm
Diameter (English) 1 in.
Diameter (Metric) 25.4 mm