missing translation for 'onlineSavingsMsg'
Learn More

Decyl Methyl Ether, TRC

Product Code. 30385624
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385624 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385624 Supplier Toronto Research Chemicals Supplier No. TRCD52537010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Decyl Methyl Ether
CAS 7289-52-3
Purity Grade Notes GC
Molecular Formula C11 H24 O
Quantity 10 g
Synonym 1-Methoxydecane, Methyl Decyl Ether, Methyl n-Decyl Ether
InChI Formula InChI=1S/C11H24O/c1-3-4-5-6-7-8-9-10-11-12-2/h3-11H2,1-2H3
SMILES CCCCCCCCCCOC
IUPAC Name 1-methoxydecane
Molecular Weight (g/mol) 172.31
Formula Weight 172.1827
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.