missing translation for 'onlineSavingsMsg'
Learn More

Decursin, TRC

Product Code. 30359137
Change view
Click to view available options
Quantity:
1 mg
25 mg
Unit Size:
1mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359137 1 mg 1mg
30363485 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30359137 Supplier Toronto Research Chemicals Supplier No. TRCD2269181MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5928-25-6
Molecular Formula C19 H20 O5
Molecular Weight (g/mol) 328.36
Synonym (7S)-7,8-Dihydro-8,8-dimethyl-2-oxo-2H,6H-benzo[1,2-b:5,4-b']dipyran-7-yl 3-Methyl-2-Butenoic Acid Ester, (S)-7,8-Dihydro-8,8-dimethyl-2-oxo-2H,6H-benzo[1,2-b:5,4-b']dipyran-7-yl 3-Methyl-2-Butenoic Acid Ester, (+)- 7,8-Dihydro-7-hydroxy-8,8-dimethyl-2H,6H-benzo[1,2-b:5,4-b']dipyran-2-one Ester with 3-methyl-crontonic Acid, 2H,6H-Benzo[1,2-b:5,4-b']dipyran-2-butenoic Acid Deriv., (+)-Decursin
IUPAC Name [(3S)-2,2-dimethyl-8-oxo-3,4-dihydropyrano[3,2-g]chromen-3-yl] 3-methylbut-2-enoate
SMILES CC(=CC(=O)O[C@H]1Cc2cc3C=CC(=O)Oc3cc2OC1(C)C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C19H20O5/c1-11(2)7-18(21)23-16-9-13-8-12-5-6-17(20)22-14(12)10-15(13)24-19(16,3)4/h5-8,10,16H,9H2,1-4H3/t16-/m0/s1
Formula Weight 328.1311
Chemical Name or Material Decursin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.