Learn More
Borer Chemie™ deconex 41 LAB CLEAR
Product Code. 17754049
Click to view available options
Quantity:
1 x 5 kg
1 x 10 kg
Unit Size:
10kg
5kg
Product Code. 17754049
Brand: Borer Chemie™ 503715.00KK5R
Neutralizes alkali residues, prevents limescale deposits in the washer and stains on the cleaned items. deconex 41 LAB CLEAR is phosphate-free and accelerates the drying of the items thanks to its wetting and clear rinsing properties.
  • Acid rinsing agent for automated use
  • Extremely economical, typical concentration 1 mL/L
  • Phosphate-free
  • Accelerates the drying of the cleaned items
  • Provides a beautiful shine
  • Technical support by the manufacturer
Specifications
Form Liquid
pH ∼2.6 (1% Solution)
Composition Organic acids, surfactants
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 5 kg
Volume (English) 1.2 gal.
Volume (Metric) 4.6 L
Weight (English) 11 lb.
Weight (Metric) 5 kg
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • deconex 41 LAB CLEAR
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide