Learn More
Borer Chemie™ deconex 26 PLUS
Click to view available options
Quantity:
1 x 7 kg
1 x 12 kg
1 x 35 kg
Unit Size:
12kg
35kg
7kg
Description
- Neutralizer and cleaner for automated use
- Economical because highly concentrated, use 2 mL/L
- Neutralizes carried-over alkaline residues, reduces rinsing phases
- Reduces the consumption of rinsing water
- Without surfactants, rinses off without leaving residues
- Also as pre-rinse agent against acid-soluble deposits
- Technical support by the manufacturer
Specifications
Specifications
| Form | Liquid |
| pH | ∼1.8 (1% Solution) |
| Composition | Inorganic acid |
| Concentration | Concentrate |
| Container Type | Canister |
| Content And Storage | 5°C to 25°C |
| Quantity | 1 x 35 kg |
| Volume (English) | 6.6 gal. |
| Volume (Metric) | 24.8 L |
| Weight (English) | 77 lb. |
| Show More |