Learn More
Borer Chemie™ deconex 25 ORGANACID
Product Code. p-7227854
Click to view available options
Quantity:
1 x 5 kg
1 x 10 kg
1 x 25 kg
Unit Size:
10kg
25kg
5kg
Product Code. 15410380
Brand: Borer Chemie™ 502100.00KK5R
Edge
Cleaner and neutralizer for laboratory washers. Shortens rinsing phases and reduces water consumption. deconex 25 ORGANACID is phosphate-free and suitable for environmental and water laboratories. Pre-rinsing agent against acid-soluble residues.
  • Neutralizer and cleaner for automated use
  • Ecological solution without phosphates and surfactants
  • Neutralizes carried-over alkaline residues, reduces rinsing phases
  • Reduces the consumption of rinsing water
  • Typical application concentration: 3 mL/L
  • Excellent material compatibility, also for aluminium
  • Can be rinsed off without leaving residues
  • Also as pre-rinsing agent against acid-soluble deposits
  • Technical support by the manufacturer
Specifications
Form Liquid
pH ∼2.6 (1% Solution)
Composition Organic acid
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 5 kg
Volume (English) 1.1 gal.
Volume (Metric) 4.3 L
Weight (English) 11 lb.
Weight (Metric) 5 kg
Show More Show Less
compliance-icons
Product Identifier
  • deconex 25 ORGANACID
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : citric acid