Learn More
Borer Chemie™ deconex 24 LIQ
Product Code. 10195443
Change view
Click to view available options
Quantity:
1 x 7 kg
1 x 14 kg
1 x 35 kg
Unit Size:
14kg
35kg
7kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10195443 1 x 7 kg 7kg
10430581 1 x 14 kg 14kg
10582101 1 x 35 kg 35kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10195443 Supplier Borer Chemie™ Supplier No. 502000.00KK5M
Liquid cleaner, for automated cleaning of laboratory glassware and utensils. For highest demands and stubborn residues, deconex 24 LIQ is suitable. An acidic rinse is recommended. Contains active chlorine as a cleaning booster.
  • Concentrate for automated cleaning
  • High yield concentrate, with active chlorine
  • Typical application concentration: 3-5 g/L
  • Can be rinsed off without leaving residues
  • For consistent and flawless results
  • Wide cleaning spectrum
  • Without EDTA, NTA and surfactants
  • Acid rinse with deconex cleaner recommended
  • Technical support by the manufacturer
Specifications
Form Liquid
pH ∼12.5 (1% Solution)
Composition Alkalis, complexing agents, corrosion inhibitor, sequestering agent, inorganic chlorine donor
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 7 kg
Volume (English) 1.2 gal.
Volume (Metric) 4.7 L
Weight (English) 15.4 lb.
Weight (Metric) 7 kg
Show More Show Less
compliance-icons
Product Identifier
  • deconex 24 LIQ
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide,tetrapotassium pyrophosphate, dipotassium trioxosilicate, sodium hypochlorite