Learn More
Fisherbrand™ Decapping Heads for Pneumatic Hand-held Crimping Tool
Product Code. 11889440 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
11 mm
13 mm
20 mm
32 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11889440 32 mm Each
11854970 13 mm Each
11854890 11 mm Each
11885050 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11889440
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with Pneumatic Hand-held Crimping Tool
  • Decapping Head
  • Pneumatic
  • Steel
TRUSTED_SUSTAINABILITY
Specifications
Description 32mm Decapping Head
Type Decapping Head
For Use With (Equipment) Aluminum Cap
Size 32 mm