missing translation for 'onlineSavingsMsg'
Learn More

Debio 0932, MedChemExpress

Código de producto. 16451867
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
missing translation for 'unitSize'
100mg
10mg
200mg
50mg
5mg
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16451867

missing translation for 'mfr': Medchem Express HY13469

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

MedChemExpress Debio 0932 (CUDC-305) is an orally active HSP90 inhibitor, with IC50s of 100 and 103 nM for HSP90α and HSP90β, respectively.

Debio 0932 (CUDC-305) is an orally active HSP90 inhibitor, with IC50s of 100 and 103 nM for HSP90α and HSP90β, respectively.

Especificaciones

Chemical Name or Material Debio 0932
CAS 1061318-81-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H30N6O2S
Quantity 50 mg
Synonym CUDC-305
Solubility Information DMSO : 75 mg/mL (169.46 mM; Need ultrasonic)
SMILES NC1=NC=CC2=C1N=C(SC3=C(N(C)C)C=C(OCO4)C4=C3)N2CCNCC(C)(C)C
Molecular Weight (g/mol) 442.58
Formula Weight 442.58
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.