Learn More
Debio 0932, MedChemExpress
Product Code. 16421086
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16421086
Brand: Medchem Express HY13469
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Debio 0932 (CUDC-305) is an orally active HSP90 inhibitor, with IC50s of 100 and 103 nM for HSP90α and HSP90β, respectively.
Debio 0932 (CUDC-305) is an orally active HSP90 inhibitor, with IC50s of 100 and 103 nM for HSP90α and HSP90β, respectively.
Specifications
Chemical Name or Material Debio 0932
CAS 1061318-81-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H30N6O2S
Quantity 200 mg
Synonym CUDC-305
Solubility Information DMSO : 75 mg/mL (169.46 mM; Need ultrasonic)
SMILES NC1=NC=CC2=C1N=C(SC3=C(N(C)C)C=C(OCO4)C4=C3)N2CCNCC(C)(C)C
Molecular Weight (g/mol) 442.58
Formula Weight 442.58
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death