missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCUN1D4 Polyclonal Antibody

Product Code. 15775571 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15775571 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15775571 Supplier Invitrogen™ Supplier No. PA557681

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FNKVMPPRKK RRPASGDDLS AKKSRHDSMY RKYDSTRIKT EEEAFSSKRC LEWFYE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

DCUN1D4 contains 1 DCUN1 domain. The exact function of DCUN1D4 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCUN1D4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCUN1D4
Gene Accession No. Q8CCA0, Q92564
Gene Alias AI836376; DCN1, defective in cullin neddylation 1, domain containing 4; DCN1, defective in cullin neddylation 1, domain containing 4 (S. cerevisiae); DCN1-like protein 4; DCUN1 domain-containing protein 4; Dcun1d4; defective in cullin neddylation 1 domain containing 4; defective in cullin neddylation protein 1-like protein 4; KIAA0276; RGD1310422; Unknown (protein for MGC:134196)
Gene Symbols DCUN1D4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DCUN1D4. Recombinant protein control fragment (Product #RP-96617).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100737, 23142, 360928
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.