Learn More
VACUUBRAND™ DCP 3000 + MPT Vacuum Gauge Set
Click to view available options
Dimensions (L x W x H):
144 x 124 x 114 mm
Unit Size:
Each
Description
- Ready for operation, with wall-plug power supply and manual.
- For the range from atmospheric pressure down to high vacuum (5 x 10-9mbar)
- Kit includes combined thermal conductivity / cold cathode (Pirani / Penning) vacuum sensor MPT.
- Can be used for relative pressure measurements (using VSK 3000 vacuum sensors as a reference) and as a data logger for up to 32,000 measurement values.
- Uniform plug and socket connections make the system flexible and allow extension of the lines up to 30m.
- Components connected to the gauges and controllers are automatically recognized, configured and supervised.
- For high vacuum measurements, the DCP 3000 can be connected with the combined Penning/Pirani gauge head MPT.
Specifications
Specifications
| Description | 100-230V, 50-60Hz |
| Dimensions (L x W x H) | 144 x 124 x 114 mm |
| Height (Metric) | 114 mm |
| Includes | Stainless steel tubing KF DN 25, Vent valve VBM-B, Vacuum sensor VSK 3000, Vacuum sensor VSP 3000, Vacuum sensor MPT 200 |
| Length (Metric) | 144 mm |
| Operating Temperature (Metric) | 10°C to 40°C |
| Weight (Metric) | 0.44 kg |
| Accuracy | 1 x 10–8 - 2 x 10–3 mbar: ±25%; 2 x 10–3 - |
| Product Type | Vacuum Gauge Set |