Learn More
Thermo Scientific™ Data Output Accessories for Environmental Chambers and Incubators
Shop All Thermo Scientific Products
Click to view available options
Model:
Chart Paper
Dual Pen
Single Pen
Type:
7 Day Chart Recorder
Chart Paper
Installation:
Customer Installed
Factory Installed
Unit Size:
Each
Description
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Specifications
Specifications
| Description | Single Pen, 6 in. 7 Day Chart Recorders; 0° to 60°C (32° to 140°F) |
| For Use With (Equipment) | Environmental Chamber (3948 and 3907) |
| Lighting | LED |
| Temperature Range (Metric) | 0°C to 60°C |
| Type | 7 Day Chart Recorder |
| Installation | Customer Installed |
| Model | Single Pen |