missing translation for 'onlineSavingsMsg'
Learn More

Dapoxetine hydrochloride, MedChemExpress

Product Code. 16454327
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16454327

Brand: Medchem Express HYB0304A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dapoxetine (LY-210448) hydrochloride is an orally active and selective serotonin reuptake inhibitor (SSRI). Dapoxetine hydrochloride can be used for the research of premature ejaculation (PE).

Dapoxetine (LY-210448) hydrochloride is an orally active and selective serotonin reuptake inhibitor (SSRI). Dapoxetine hydrochloride can be used for the research of premature ejaculation (PE).

Specifications

Chemical Name or Material Dapoxetine hydrochloride
CAS 129938-20-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H24ClNO
Quantity 50 mg
Synonym LY-210448 hydrochloride
Solubility Information DMSO : ≥ 32 mg/mL (93.60 mM)
SMILES CN(C)[C@H](C1=CC=CC=C1)CCOC2=C3C(C=CC=C3)=CC=C2.Cl
Molecular Weight (g/mol) 341.87
Formula Weight 341.87
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.