Learn More
Dapansutrile, MedChemExpress
Product Code. 16407438
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 16407438
Brand: Medchem Express HY17629
MedChemExpress Dapansutrile is a potent, orally active and selective NLRP3 inflammasome inhibitor. Dapansutrile has anti-inflammatory activity and decreases immune factor levels. Dapansutrile can be used for research of inflammatory diseases.
Dapansutrile is a potent, orally active and selective NLRP3 inflammasome inhibitor. Dapansutrile has anti-inflammatory activity and decreases immune factor levels. Dapansutrile can be used for research of inflammatory diseases.
Specifications
Chemical Name or Material Dapansutrile
Physical Form Solid
CAS 54863-37-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C4H7NO2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 125 mg/mL (938.65 mM) ∣H2O : 36.67 mg/mL (275.36 mM; Need ultrasonic)
SMILES N#CCCS(=O)(C)=O
Molecular Weight (g/mol) 133.17
Formula Weight 133.17
Percent Purity 98.0%
Grade Research